siRNA Details
| Gene Symbol: MYLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033118.3 | NP_149109.1 | 2, 3 | 157 | ||
| GenBank mRNA | AF325549.1 | - | 1, 2 | 55 | ||
| AJ272502.1 | - | 2, 3 | 163 | |||
| BC069627.1 | - | 1, 2 | 55 | 7262446 | ||
| BC092413.1 | - | 2, 3 | 157 | 30530456 | ||
| BC127622.1 | - | 1, 2 | 55 | 40085085 | ||
| BX647635.1 | - | 2, 3 | 140 | |||

MYLK2
Genomic Map