siRNA Details
| Gene Symbol: TUBGCP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020461.3 | NP_065194.2 | 1 | 905 | ||
| GenBank mRNA | AB051456.1 | - | 1 | 299 | ||
| AF272887.1 | - | 1 | 498 | |||
| AK310419.1 | - | 1 | 939 | |||
| AL832617.2 | - | 1 | 926 | |||
| BC144488.1 | - | 1 | 497 | 9053015 | ||
| BC144489.1 | - | 1 | 497 | |||
| BC150613.1 | - | 1 | 497 | 9056983 | ||
| BC151209.1 | - | 1 | 887 | 40147276 | ||

TUBGCP6
Genomic Map