siRNA Details
| Gene Symbol: DOCK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271999.1 | NP_001258928.1 | 19 | 2237 | ||
| NM_001272000.1 | NP_001258929.1 | 19 | 2237 | |||
| NM_001272001.1 | NP_001258930.1 | 19 | 2237 | |||
| NM_033407.3 | NP_212132.2 | 19 | 2237 | |||
| XM_005271292.2 | XP_005271349.1 | 19 | 2227 | |||
| XM_011542326.2 | XP_011540628.1 | 19 | 2227 | |||
| XM_011542327.2 | XP_011540629.1 | 19 | 2227 | |||
| XM_011542328.2 | XP_011540630.1 | 19 | 2227 | |||
| XM_011542330.2 | XP_011540632.1 | 19 | 2227 | |||
| XM_017002639.1 | XP_016858128.1 | 19 | 2227 | |||
| XM_017002640.1 | XP_016858129.1 | 19 | 2227 | |||
| GenBank mRNA | AK125049.1 | - | 5 | 382 | ||
| BX485492.1 | - | 3 | 174 | |||
| DQ118679.1 | - | 16 | 1809 | |||
| DQ118680.1 | - | 16 | 1809 | |||
| DQ309763.1 | - | 16 | 1809 | |||
| DQ341187.1 | - | 19 | 2144 | |||

DOCK7
Genomic Map