siRNA Details
| Gene Symbol: ADNP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014913.3 | NP_055728.1 | 4 | 2793 | ||
| XM_005266656.4 | XP_005266713.1 | 4 | 2647 | |||
| XM_011525883.2 | XP_011524185.1 | 4 | 2507 | |||
| GenBank mRNA | AB020670.1 | - | 4 | 2523 | ||
| AK022688.1 | - | 1 | 1311 | |||
| AK292566.1 | - | 4 | 2562 | |||
| BC071589.1 | - | 4 | 2793 | 30333923 | ||

ADNP2
Genomic Map