siRNA Details
Gene Symbol: JMJD7-PLA2G4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198588.1 | NP_001185517.1 | 9 | 974 | ||
NM_005090.3 | NP_005081.1 | 9 | 974 | |||
GenBank mRNA | AB209150.1 | - | 9 | 4442 | ||
AF065215.1 | - | 9 | 949 | |||
AF065216.1 | - | 9 | 4279 | |||
AF121908.1 | - | 9 | 941 | |||
DQ523799.1 | - | 9 | 942 | |||
DQ523800.1 | - | 9 | 942 |
Gene Symbol: PLA2G4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114633.1 | NP_001108105.1 | 4 | 342 | ||
GenBank mRNA | AK299419.1 | - | 4 | 308 | ||
AK310651.1 | - | 4 | 342 | |||
DA250544.1 | - | 1 | 23 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
