siRNA Details
Gene Symbol: S1PR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003775.3 | NP_003766.1 | 1 | 294 | ||
GenBank mRNA | AJ000479.1 | - | 1 | 261 | ||
BC014970.1 | - | 1 | 264 | 4849349 | ||
FJ200495.1 | - | 2 | 101 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
