siRNA Details
| Gene Symbol: ZNF502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134440.1 | NP_001127912.1 | 4 | 1584 | ||
| NM_001134441.1 | NP_001127913.1 | 3 | 1490 | |||
| NM_001134442.1 | NP_001127914.1 | 3 | 1486 | |||
| NM_001282880.1 | NP_001269809.1 | 5 | 1707 | |||
| NM_033210.4 | NP_149987.2 | 4 | 1588 | |||
| XM_017007440.1 | XP_016862929.1 | 2 | 2826 | |||
| GenBank mRNA | AK022577.1 | - | 1 | 918 | ||
| AK027761.1 | - | 3 | 1482 | |||
| BC028377.2 | - | 4 | 1585 | 4822224 | ||

ZNF502
Genomic Map