siRNA Details
| Gene Symbol: ZNF235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004234.4 | NP_004225.3 | 5 | 822 | ||
| XM_005259412.4 | XP_005259469.1 | 5 | 4878 | |||
| GenBank mRNA | AK226126.1 | - | 1 | 1851 | ||
| AK300317.1 | - | 5 | 822 | |||
| AK315242.1 | - | 5 | 797 | |||
| BC117215.1 | - | 5 | 795 | 40125766 | ||
| BC143482.1 | - | 5 | 795 | |||

ZNF235
Genomic Map