siRNA Details
| Gene Symbol: HS6ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004807.2 | NP_004798.3 | 2 | 1574 | ||
| GenBank mRNA | AB006179.1 | - | 2 | 1648 | ||
| BC001196.1 | - | 1 | 823 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS6ST1| Gene Symbol: HS6ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004807.2 | NP_004798.3 | 2 | 1574 | ||
| GenBank mRNA | AB006179.1 | - | 2 | 1648 | ||
| BC001196.1 | - | 1 | 823 | |||