siRNA Details
| Gene Symbol: CABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033677.1 | NP_001028849.1 | 6 | 1347 | ||
| NM_004276.4 | NP_004267.2 | 6 | 802 | |||
| NM_031205.3 | NP_112482.1 | 7 | 982 | |||
| XM_017020237.1 | XP_016875726.1 | 6 | 1549 | |||
| GenBank mRNA | BC030201.1 | - | 6 | 1347 | 5200625 | |
| BQ188859.1 | - | 2 | 237 | |||

CABP1
Genomic Map