siRNA Details
| Gene Symbol: ATG14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014924.4 | NP_055739.2 | 8 | 1051 | ||
| XM_011536563.2 | XP_011534865.1 | 6 | 755 | |||
| GenBank mRNA | AK131251.1 | - | 8 | 1051 | ||
| AK292810.1 | - | 8 | 1023 | |||
| BC109089.1 | - | 8 | 1036 | 40034573 | ||
| BC109090.1 | - | 8 | 1036 | 40034574 | ||

ATG14
Genomic Map