siRNA Details
| Gene Symbol: SLITRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318810.1 | NP_001305739.1 | 2 | 836 | ||
| NM_001318811.1 | NP_001305740.1 | 2 | 489 | |||
| NM_014926.3 | NP_055741.2 | 2 | 674 | |||
| GenBank mRNA | AB020655.1 | - | 2 | 503 | ||
| AK290024.1 | - | 2 | 836 | |||
| BC114621.1 | - | 2 | 488 | 40036558 | ||
| DA233757.1 | - | 2 | 489 | |||

SLITRK3
Genomic Map