siRNA Details
| Gene Symbol: ZNF510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001314059.1 | NP_001300988.1 | 6 | 1662 | ||
| NM_001314060.1 | NP_001300989.1 | 5 | 1214 | |||
| NM_014930.2 | NP_055745.1 | 6 | 1273 | |||
| XM_005251808.2 | XP_005251865.1 | 6 | 1260 | |||
| XM_011518393.2 | XP_011516695.1 | 6 | 1718 | |||
| XM_017014483.1 | XP_016869972.1 | 7 | 1325 | |||
| GenBank mRNA | AB023189.1 | - | 5 | 1190 | ||
| AK292600.1 | - | 6 | 1243 | |||
| BC036676.1 | - | 6 | 1660 | 4792684 | ||
| BC058022.1 | - | 6 | 1237 | |||
| BC068587.1 | - | 6 | 1656 | 4374918 | ||

ZNF510
Genomic Map