siRNA Details
| Gene Symbol: UBAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016172.2 | NP_057256.2 | 9, 10 | 1314 | ||
| GenBank mRNA | AF038200.1 | - | 8, 9 | 879 | ||
| AF068195.1 | - | 8, 9 | 941 | |||
| AF176796.1 | - | 9, 10 | 1284 | |||
| BC004967.1 | - | 9, 10 | 1314 | 3535682 | ||
| BC011822.1 | - | 9, 10 | 1140 | 3688526 | ||
| BX647463.1 | - | 2, 3 | 5367 | |||

UBAC1
Genomic Map