siRNA Details
| Gene Symbol: SH2D1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053282.4 | NP_444512.2 | 3 | 470 | ||
| GenBank mRNA | AF256653.1 | - | 3 | 348 | ||
| AF403479.1 | - | 3 | 348 | |||
| AF484964.1 | - | 3 | 446 | |||
| AK097162.1 | - | 3 | 397 | |||
| AK314743.1 | - | 3 | 417 | |||
| BC022407.1 | - | 3 | 458 | 4717041 | ||

SH2D1B
Genomic Map