siRNA Details
| Gene Symbol: BTBD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318189.1 | NP_001305118.1 | 10 | 1143 | ||
| NM_144587.3 | NP_653188.2 | 10 | 1140 | |||
| XM_011539240.2 | XP_011537542.1 | 9 | 1549 | |||
| XM_011539241.2 | XP_011537543.1 | 8 | 1492 | |||
| XM_017015636.1 | XP_016871125.1 | 9 | 2223 | |||
| XM_017015637.1 | XP_016871126.1 | 6 | 1848 | |||
| GenBank mRNA | AK058088.1 | - | 10 | 1130 | ||
| BC109261.1 | - | 10 | 1114 | 40025404 | ||
| BC109262.1 | - | 10 | 1114 | 40025414 | ||

BTBD16
Genomic Map