siRNA Details
| Gene Symbol: OR56A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005179.2 | NP_001005179.2 | 1 | 801 | ||
| GenBank mRNA | BC136986.1 | - | 1 | 847 | 9020978 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR56A4| Gene Symbol: OR56A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005179.2 | NP_001005179.2 | 1 | 801 | ||
| GenBank mRNA | BC136986.1 | - | 1 | 847 | 9020978 | |
Set Membership: |
|