siRNA Details
| Gene Symbol: C12orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152318.2 | NP_689531.2 | 3 | 350 | ||
| GenBank mRNA | BC032326.1 | - | 334 | 4385532 | ||
| BG829346.1 | - | 3 | 351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12orf45| Gene Symbol: C12orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152318.2 | NP_689531.2 | 3 | 350 | ||
| GenBank mRNA | BC032326.1 | - | 334 | 4385532 | ||
| BG829346.1 | - | 3 | 351 | |||