siRNA Details
| Gene Symbol: UBALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145253.2 | NP_660296.1 | 3 | 1255 | ||
| XM_017022937.1 | XP_016878426.1 | 3 | 1154 | |||
| GenBank mRNA | AF447881.1 | - | 1 | 2482 | ||
| AK056747.1 | - | 2 | 1912 | |||
| BC008681.1 | - | 3 | 1210 | |||
| BC012018.1 | - | 3 | 1177 | 4477559 | ||
| BC025327.1 | - | 3 | 1198 | 3614122 | ||
| BC062534.1 | - | 3 | 1255 | 4620605 | ||

UBALD1
Genomic Map