siRNA Details
| Gene Symbol: OR7D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005191.2 | NP_001005191.1 | 1 | 784 | ||
| GenBank mRNA | BC137147.1 | - | 1 | 784 | 9021144 | |
| BC137162.1 | - | 1 | 784 | 9021159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR7D4| Gene Symbol: OR7D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005191.2 | NP_001005191.1 | 1 | 784 | ||
| GenBank mRNA | BC137147.1 | - | 1 | 784 | 9021144 | |
| BC137162.1 | - | 1 | 784 | 9021159 | ||