siRNA Details
| Gene Symbol: JOSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270639.1 | NP_001257568.1 | 5 | 973 | ||
| NM_001270640.1 | NP_001257569.1 | 5 | 970 | |||
| NM_001270641.1 | NP_001257570.1 | 4 | 847 | |||
| NM_001270686.1 | NP_001257615.1 | 5 | 816 | |||
| NM_138334.3 | NP_612207.1 | 5 | 819 | |||
| XM_011526434.2 | XP_011524736.1 | 5 | 813 | |||
| GenBank mRNA | AF247787.1 | - | 4 | 748 | ||
| AK131052.1 | - | 4 | 712 | |||
| BC051380.1 | - | 5 | 777 | |||
| BC062416.1 | - | 5 | 796 | 6503623 | ||

JOSD2
Genomic Map