siRNA Details
| Gene Symbol: ZNF428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182498.3 | NP_872304.2 | 3 | 1307 | ||
| GenBank mRNA | BC045799.1 | - | 3 | 1048 | 4813427 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF428| Gene Symbol: ZNF428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182498.3 | NP_872304.2 | 3 | 1307 | ||
| GenBank mRNA | BC045799.1 | - | 3 | 1048 | 4813427 | |