siRNA Details
| Gene Symbol: JSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144616.3 | NP_653217.1 | 7 | 702 | ||
| XM_017026285.1 | XP_016881774.1 | 7 | 1117 | |||
| XM_017026286.1 | XP_016881775.1 | 7 | 1096 | |||
| XM_017026287.1 | XP_016881776.1 | 7 | 699 | |||
| GenBank mRNA | AK056978.1 | - | 7 | 696 | ||
| BC021201.2 | - | 6 | 666 | 4106867 | ||

JSRP1
Genomic Map