siRNA Details
Gene Symbol: C1orf122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142726.1 | NP_001136198.1 | 3 | 1144 | ||
NM_198446.2 | NP_940848.2 | 3 | 1277 | |||
GenBank mRNA | AK127381.1 | - | 3 | 781 | ||
AY517499.1 | - | 3 | 932 | |||
BC016402.1 | - | 2 | 2190 | |||
BC057819.1 | - | 3 | 782 | 4792982 | ||
BC107856.1 | - | 2 | 892 |
