siRNA Details
| Gene Symbol: KLHDC7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152375.2 | NP_689588.2 | 1 | 837 | ||
| GenBank mRNA | AK096072.1 | - | 1 | 395 | ||
| BC101498.1 | - | 1 | 203 | |||
| BC101500.1 | - | 1 | 203 | |||
| BC151205.1 | - | 1 | 596 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHDC7A| Gene Symbol: KLHDC7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152375.2 | NP_689588.2 | 1 | 837 | ||
| GenBank mRNA | AK096072.1 | - | 1 | 395 | ||
| BC101498.1 | - | 1 | 203 | |||
| BC101500.1 | - | 1 | 203 | |||
| BC151205.1 | - | 1 | 596 | |||