siRNA Details
| Gene Symbol: C2orf73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100396.1 | NP_001093866.1 | 4, 5 | 554 | ||
| XM_005264133.1 | XP_005264190.1 | 3, 4 | 346 | |||
| XM_005264134.3 | XP_005264191.1 | 3, 4 | 429 | |||
| XM_011532510.2 | XP_011530812.1 | 4, 5 | 624 | |||
| XM_011532513.2 | XP_011530815.1 | 3, 4 | 527 | |||
| XM_017003323.1 | XP_016858812.1 | 4, 5 | 449 | |||
| GenBank mRNA | AK097617.1 | - | 3, 4 | 346 | ||
| BC031669.1 | - | 4, 5 | 517 | |||
| BC126244.1 | - | 3, 4 | 315 | 8991960 | ||
| BC126246.1 | - | 3, 4 | 315 | 8991962 | ||
| BC144192.1 | - | 4, 5 | 349 | 9052712 | ||

C2orf73
Genomic Map