siRNA Details
| Gene Symbol: C2orf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017927.3 | NP_001017927.2 | 3 | 691 | ||
| NM_001322329.1 | NP_001309258.1 | 3 | 612 | |||
| NM_001322330.1 | NP_001309259.1 | 2 | 488 | |||
| NM_001322331.1 | NP_001309260.1 | 2 | 141 | |||
| NM_001322332.1 | NP_001309261.1 | 2 | 141 | |||
| XM_017003354.1 | XP_016858843.1 | 2 | 190 | |||
| GenBank mRNA | BC047365.1 | - | 3 | 612 | 5189916 | |
| BC062434.1 | - | 1 | 123 | |||
| BC105805.1 | - | 2 | 145 | 6496685 | ||
| BC126395.1 | - | 2 | 428 | 8992111 | ||
| BC126397.1 | - | 3 | 631 | 8992113 | ||
| BC144012.1 | - | 3 | 631 | 9052531 | ||
| BC171737.1 | - | 2 | 428 | 9052532 | ||

C2orf76
Genomic Map