siRNA Details
| Gene Symbol: DNAJC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190233.1 | NP_001177162.1 | 5, 6 | 471 | ||
| NM_145261.3 | NP_660304.1 | 5, 6 | 459 | |||
| NR_033721.1 | 4, 5 | 407 | ||||
| NR_033722.1 | 4, 5 | 379 | ||||
| NR_033723.1 | 5, 6 | 404 | ||||
| NR_046073.1 | 2, 3 | 253 | ||||
| GenBank mRNA | AK311765.1 | - | 5, 6 | 387 | ||
| BC073989.1 | - | 5, 6 | 404 | 5760865 | ||
| BF666599.1 | - | 5, 6 | 421 | |||
| BG492744.1 | - | 4, 5 | 344 | |||
| BM701756.1 | - | 4, 5 | 332 | |||
| BM702203.1 | - | 4, 5 | 332 | |||
| BQ438379.1 | - | 2, 3 | 190 | |||
| CB052107.1 | - | 5, 6 | 342 | |||
| CB054787.1 | - | 2, 3 | 175 | |||
| CB159371.1 | - | 5, 6 | 471 | |||
| KU178861.1 | - | 4, 5 | 287 | |||
| KU178862.1 | - | 3, 4 | 207 | |||

DNAJC19
Genomic Map