siRNA Details
| Gene Symbol: SCLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144643.3 | NP_653244.2 | 12 | 1519 | ||
| XM_017007716.1 | XP_016863205.1 | 12 | 1519 | |||
| XM_017007717.1 | XP_016863206.1 | 12 | 1519 | |||
| XM_017007718.1 | XP_016863207.1 | 12 | 1519 | |||
| XM_017007719.1 | XP_016863208.1 | 12 | 1040 | |||
| XM_017007720.1 | XP_016863209.1 | 13 | 1431 | |||
| XM_017007721.1 | XP_016863210.1 | 6 | 633 | |||
| GenBank mRNA | AK055217.1 | - | 12 | 1519 | ||
| AK093361.1 | - | 5 | 482 | |||
| AK122852.1 | - | 2 | 2142 | |||

SCLT1
Genomic Map