siRNA Details
| Gene Symbol: WBSCR28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182504.3 | NP_872310.2 | 1 | 92 | ||
| GenBank mRNA | AY372053.1 | - | 1 | 97 | ||
| AY372054.1 | - | 1 | 19 | |||
| BC030643.2 | - | 1 | 92 | 4824662 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WBSCR28| Gene Symbol: WBSCR28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182504.3 | NP_872310.2 | 1 | 92 | ||
| GenBank mRNA | AY372053.1 | - | 1 | 97 | ||
| AY372054.1 | - | 1 | 19 | |||
| BC030643.2 | - | 1 | 92 | 4824662 | ||