siRNA Details
| Gene Symbol: KRT80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081492.1 | NP_001074961.1 | 2 | 527 | ||
| NM_182507.2 | NP_872313.2 | 2 | 527 | |||
| XM_005268676.3 | XP_005268733.1 | 1 | 328 | |||
| GenBank mRNA | AJ717743.1 | - | 2 | 481 | ||
| BC065180.1 | - | 2 | 527 | 6573884 | ||
| BX537567.1 | - | 1 | 291 | |||

KRT80
Genomic Map