siRNA Details
| Gene Symbol: C16orf92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109659.1 | NP_001103129.1 | 2 | 120 | ||
| NM_001109660.1 | NP_001103130.1 | 2 | 200 | |||
| GenBank mRNA | AK058133.1 | - | 2 | 200 | ||
| BC101433.1 | - | 2 | 120 | 40021839 | ||
| BC101434.1 | - | 2 | 120 | 40021838 | ||
| BC101435.1 | - | 2 | 120 | 40021831 | ||
| BC101436.1 | - | 2 | 200 | 40021833 | ||
| BC127955.1 | - | 2 | 200 | 40023544 | ||

C16orf92
Genomic Map