siRNA Details
| Gene Symbol: TRAPPC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014939.3 | NP_055754.2 | 7 | 1472 | ||
| XM_005258233.2 | XP_005258290.1 | 7 | 1565 | |||
| XM_005258234.2 | XP_005258291.1 | 7 | 1565 | |||
| XM_005258235.3 | XP_005258292.1 | 2 | 346 | |||
| XM_006722420.3 | XP_006722483.1 | 7 | 1565 | |||
| XM_011525884.2 | XP_011524186.1 | 7 | 1565 | |||
| XM_017025615.1 | XP_016881104.1 | 7 | 1064 | |||
| XM_017025616.1 | XP_016881105.1 | 7 | 1562 | |||
| XM_017025617.1 | XP_016881106.1 | 7 | 1562 | |||
| GenBank mRNA | AB023229.1 | - | 7 | 1165 | ||
| AK292537.1 | - | 7 | 1451 | |||
| AL136749.1 | - | 8 | 1259 | |||
| BC059380.1 | - | 7 | 1451 | |||
| BC127109.1 | - | 7 | 1026 | 40124744 | ||
| BC127110.1 | - | 7 | 1026 | 40124745 | ||

TRAPPC8
Genomic Map