siRNA Details
| Gene Symbol: PPM1N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080401.1 | NP_001073870.1 | 5 | 1447 | ||
| XM_011526474.2 | XP_011524776.1 | 4 | 1895 | |||
| GenBank mRNA | AK097444.1 | - | 4 | 1850 | ||
| BC062452.1 | - | 5 | 625 | |||

PPM1N
Genomic Map