siRNA Details
Gene Symbol: PPM1N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080401.1 | NP_001073870.1 | 5 | 1561 | ||
XM_011526474.2 | XP_011524776.1 | 4 | 2009 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PPM1N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080401.1 | NP_001073870.1 | 5 | 1561 | ||
XM_011526474.2 | XP_011524776.1 | 4 | 2009 |