siRNA Details
| Gene Symbol: DPY19L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172774.1 | NP_001166245.1 | 6, 7 | 788 | ||
| NM_207325.2 | NP_997208.2 | 6, 7 | 814 | |||
| XM_011526526.2 | XP_011524828.1 | 6, 7 | 769 | |||
| XM_017026366.1 | XP_016881855.1 | 6, 7 | 805 | |||
| XM_017026367.1 | XP_016881856.1 | 6, 7 | 772 | |||
| GenBank mRNA | AK126757.1 | - | 6, 7 | 805 | ||
| AK129497.1 | - | 6, 7 | 769 | |||
| BC131725.1 | - | 5, 6 | 636 | 40123868 | ||

DPY19L3
Genomic Map