siRNA Details
| Gene Symbol: RNF187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010858.2 | NP_001010858.2 | 4 | 977 | ||
| GenBank mRNA | BC008022.1 | - | 2 | 182 | ||
| BC012758.2 | - | 4 | 951 | |||
| BC064481.1 | - | 4 | 511 | |||
| BC080645.1 | - | 4 | 511 | |||

RNF187
Genomic Map