siRNA Details
| Gene Symbol: CXorf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025265.2 | NP_001020436.1 | 6 | 734 | ||
| NR_033212.1 | 6 | 715 | ||||
| XM_005262244.3 | XP_005262301.1 | 5 | 850 | |||
| GenBank mRNA | BC144434.1 | - | 6 | 532 | ||
| DB523273.1 | - | 5 | 418 | |||

CXorf65
Genomic Map