siRNA Details
| Gene Symbol: SAMD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010860.1 | NP_001010860.1 | 1 | 1092 | ||
| GenBank mRNA | AK093282.1 | - | 1 | 1108 | ||
| AK292571.1 | - | 1 | 1378 | |||
| BC104834.1 | - | 1 | 1112 | 8143837 | ||
| BC104836.1 | - | 1 | 1112 | 8143839 | ||

SAMD15
Genomic Map