siRNA Details
| Gene Symbol: KRT28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181535.3 | NP_853513.2 | 2 | 535 | ||
| GenBank mRNA | AJ564207.1 | - | 2 | 508 | ||
| AK129827.1 | - | 2 | 535 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT28| Gene Symbol: KRT28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181535.3 | NP_853513.2 | 2 | 535 | ||
| GenBank mRNA | AJ564207.1 | - | 2 | 508 | ||
| AK129827.1 | - | 2 | 535 | |||