siRNA Details
| Gene Symbol: PIANP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244014.1 | NP_001230943.1 | 5 | 2257 | ||
| NM_001244015.1 | NP_001230944.1 | 6 | 1951 | |||
| NM_153685.3 | NP_710152.1 | 6 | 1885 | |||
| GenBank mRNA | AK056295.1 | - | 6 | 1842 | ||
| AK091596.1 | - | 6 | 1979 | |||
| AK091656.1 | - | 5 | 2057 | |||
| AK095854.1 | - | 6 | 1951 | |||
| AL831874.1 | - | 1 | 571 | |||
| AY358534.1 | - | 4 | 2041 | |||
| BC035736.1 | - | 6 | 1688 | 5730150 | ||
| BM703122.1 | - | 1 | 315 | |||

PIANP
Genomic Map