siRNA Details
| Gene Symbol: SPATA24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194296.1 | NP_919272.1 | 1 | 126 | ||
| XM_005271916.4 | XP_005271973.1 | 1 | 243 | |||
| XM_011543252.2 | XP_011541554.1 | 1 | 72 | |||
| XM_011543253.2 | XP_011541555.1 | 1 | 71 | |||
| XM_011543254.2 | XP_011541556.1 | 1 | 176 | |||
| GenBank mRNA | AK098740.1 | - | 1 | 127 | ||
| BC040037.1 | - | 1 | 106 | |||
| BC050563.1 | - | 1 | 106 | 5751687 | ||
| BM785501.1 | - | 1 | 127 | |||

SPATA24
Genomic Map