siRNA Details
| Gene Symbol: CCDC107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195200.1 | NP_001182129.1 | 4, 5 | 499 | ||
| NM_174923.2 | NP_777583.2 | 4, 5 | 499 | |||
| GenBank mRNA | BC018758.1 | - | 4, 5 | 475 | 4907976 | |
| BE903559.1 | - | 4, 5 | 488 | |||
| DQ884403.1 | - | 4, 5 | 409 | |||

CCDC107
Genomic Map