siRNA Details
| Gene Symbol: EVI2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003927.2 | NP_001003927.1 | 3 | 653 | ||
| NM_014210.3 | NP_055025.2 | 2 | 512 | |||
| GenBank mRNA | AK295311.1 | - | 3 | 588 | ||
| AK312348.1 | - | 2 | 465 | |||
| AV724051.1 | - | 2 | 433 | |||
| BC035572.1 | - | 2 | 400 | 5229926 | ||
| BG699778.1 | - | 3 | 553 | |||
| BG700097.1 | - | 3 | 610 | |||
| BI597493.1 | - | 2 | 468 | |||
| DA364537.1 | - | 2 | 512 | |||

EVI2A
Genomic Map