siRNA Details
| Gene Symbol: WDR37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014023.3 | NP_054742.2 | 3 | 388 | ||
| GenBank mRNA | AB023199.1 | - | 3 | 359 | ||
| AK128194.1 | - | 2 | 3197 | |||
| AK292591.1 | - | 3 | 351 | |||
| AL136827.1 | - | 3 | 366 | |||
| BC018044.1 | - | 3 | 417 | 4794184 | ||

WDR37
Genomic Map