siRNA Details
| Gene Symbol: JAZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175061.3 | NP_778231.2 | 4 | 681 | ||
| XM_006715656.1 | XP_006715719.1 | 5 | 557 | |||
| XM_017011831.1 | XP_016867320.1 | 4 | 449 | |||
| XM_017011832.1 | XP_016867321.1 | 4 | 381 | |||
| GenBank mRNA | AK091311.1 | - | 4 | 606 | ||
| AL832491.1 | - | 2 | 259 | |||
| AL834234.1 | - | 3 | 400 | |||
| AY372319.1 | - | 4 | 459 | |||
| BC042441.1 | - | 5 | 469 | 4831063 | ||
| BC047229.1 | - | 4 | 468 | |||

JAZF1
Genomic Map