siRNA Details
Gene Symbol: ABLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301015.1 | NP_001287944.1 | 9, 10 | 1508 | ||
NM_001301018.1 | NP_001287947.1 | 9, 10 | 982 | |||
NM_014945.3 | NP_055760.1 | 10, 11 | 1154 | |||
XM_005268392.1 | XP_005268449.1 | 10, 11 | 1135 | |||
XM_005268393.1 | XP_005268450.1 | 10, 11 | 1135 | |||
XM_011537609.2 | XP_011535911.1 | 10, 11 | 1135 | |||
GenBank mRNA | AB020650.1 | - | 10, 11 | 1067 | ||
AK225769.1 | - | 9, 10 | 1376 | |||
AK289736.1 | - | 9, 10 | 921 | |||
DQ413174.1 | - | 9, 10 | 944 |
