siRNA Details
| Gene Symbol: RNF44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014901.4 | NP_055716.1 | 11 | 2639 | ||
| XM_005265840.2 | XP_005265897.1 | 11 | 2351 | |||
| XM_005265841.4 | XP_005265898.1 | 11 | 2596 | |||
| XM_005265842.4 | XP_005265899.1 | 11 | 2581 | |||
| XM_005265843.2 | XP_005265900.1 | 11 | 2287 | |||
| XM_005265844.3 | XP_005265901.1 | 11 | 2392 | |||
| XM_005265845.4 | XP_005265902.1 | 11 | 2530 | |||
| XM_006714831.2 | XP_006714894.1 | 11 | 2342 | |||
| XM_006714832.2 | XP_006714895.1 | 11 | 2210 | |||
| XM_011534466.2 | XP_011532768.1 | 11 | 2354 | |||
| XM_011534467.2 | XP_011532769.1 | 11 | 2585 | |||
| GenBank mRNA | AB029023.1 | - | 11 | 2524 | ||
| AK127673.1 | - | 2 | 1461 | |||
| BC039833.1 | - | 11 | 2595 | 5535911 | ||
| BC063297.1 | - | 11 | 2640 | 30349418 | ||

RNF44
Genomic Map