siRNA Details
| Gene Symbol: UBOX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267584.1 | NP_001254513.1 | 3 | 555 | ||
| NM_014948.3 | NP_055763.1 | 3 | 555 | |||
| NM_199415.2 | NP_955447.1 | 3 | 555 | |||
| GenBank mRNA | AB020667.1 | - | 3 | 523 | ||
| AK022444.1 | - | 3 | 519 | |||
| AK222570.1 | - | 3 | 527 | |||
| AK222876.1 | - | 3 | 480 | |||
| BC000515.2 | - | 3 | 475 | 2822816 | ||
| BC046122.1 | - | 3 | 515 | 5554044 | ||
| CR457089.1 | - | 2 | 369 | |||
| DA101925.1 | - | 3 | 555 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_080562, NM_001255993, NM_001255994, BC025068, AK034114, and others.

UBOX5
Genomic Map