siRNA Details
Gene Symbol: FRAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012083.2 | NP_036215.1 | 1 | 1742 | ||
GenBank mRNA | AB045118.1 | - | 1 | 1742 | ||
BC020165.1 | - | 1 | 1717 | 3629702 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FRAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012083.2 | NP_036215.1 | 1 | 1742 | ||
GenBank mRNA | AB045118.1 | - | 1 | 1742 | ||
BC020165.1 | - | 1 | 1717 | 3629702 |